Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, >-99%, CHCl3, CAS Number-67-66-3, 64-17-5, trichloromethane, trichloroform, trichlormethan, methyl trichloride, cloroformio, methenyl trichloride, formyl trichloride, methane, trichloro, chloroforme, methane trichloride, 2.5L
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR390760025
Price (MYR)
1,023.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,023.00
Add to cart
Chemical Identifiers
CAS7790-78-5
IUPAC Namecadmium(2+) dichloride
Molecular FormulaCdCl2
InChI KeyYKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<50 ppm
Lead (Pb)=<50 ppm
Appearance (Form)Crystalline powder or crystals and/or chunks
Insoluble matter=<0.005 % (in water)
Ammonium (NH4)=<50 ppm
View more
For HPLC
RUO – Research Use Only