Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, >-99%, CHCl3, CAS Number-67-66-3, 64-17-5, trichloromethane, trichloroform, trichlormethan, methyl trichloride, cloroformio, methenyl trichloride, formyl trichloride, methane, trichloro, chloroforme, methane trichloride, 2.5L
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR390760010
Price (MYR)
750.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
750.00
Add to cart
Chemical Identifiers
CAS57-57-8
IUPAC Nameoxetan-2-one
Molecular FormulaC3H4O2
InChI KeyVEZXCJBBBCKRPI-UHFFFAOYSA-N
SMILESO=C1CCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥96.0% (U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.4105-1.4155 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless
Assay (GC)≥96.0% (non-U.S. specification)
View more
For HPLC
RUO – Research Use Only