tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic Bottle
Catalog number ACR390690025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS7446-70-0
IUPAC Namealuminium(3+) trichloride
Molecular FormulaAlCl3
InChI KeyVSCWAEJMTAWNJL-UHFFFAOYSA-K
SMILES[Al+3].[Cl-].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
CommentIron: ≤50 ppm
CommentZinc: Report
CommentCopper: Report
Assay (unspecified)≥99.0%