1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR390682500
Price (MYR)
469.00
Add to cart
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
469.00
Add to cart
Chemical Identifiers
CAS822-87-7
IUPAC Name2-chlorocyclohexan-1-one
Molecular FormulaC6H9ClO
InChI KeyCCHNWURRBFGQCD-UHFFFAOYNA-N
SMILESClC1CCCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Refractive index1.4830 to 1.4850 (20°C, 589 nm)
Stabilizer0.2 % CaCO3/MgO (1:1)
Appearance (Color)Clear brown
Appearance (Form)Liquid
View more
Suitable for battery materials development.
RUO – Research Use Only