1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass Bottle
Catalog number ACR390680025
Price (MYR)
1,407.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,407.00
BOT
Add to cart
Chemical Identifiers
CAS108-48-5
IUPAC Name2,6-dimethylpyridine
Molecular FormulaC7H9N
InChI KeyOISVCGZHLKNMSJ-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4966 to 1.4986 (20°C, 589 nm)
AppearanceMay darken on storage
Water=<0.2 %
Appearance (Form)Clear liquid
View more
Suitable for battery materials development.
RUO – Research Use Only