1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
Packaging:
Glass Bottle
Catalog number ACR390680010
Price (MYR)
682.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
682.00
Add to cart
Chemical Identifiers
CAS5989-27-5
IUPAC Name(4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
Molecular FormulaC10H16
InChI KeyXMGQYMWWDOXHJM-JTQLQIEISA-N
SMILESCC(=C)[C@@H]1CCC(C)=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4710 to 1.4740 (20°C, 589 nm)
Specific optical rotation+102° to +110° (20°C, 589 nm) (C=1, CH3OH)
GC>=95 %
View more
Suitable for battery materials development.
RUO – Research Use Only