N,N-Dimethylacetamide, 99.8+%, for HPLC, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.8+%, for HPLC, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.8+%, for HPLC, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.8+%, for HPLC, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR390670025
Price (MYR)
1,484.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,484.00
Add to cart
Chemical Identifiers
CAS10233-87-1
IUPAC Namemagnesium(2+) pentahydrate phosphate
Molecular FormulaH10MgO9P
InChI KeyZHDVAZVXDZNFPS-UHFFFAOYSA-K
SMILESO.O.O.O.O.[Mg++].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)P_2O_5 33% min
For HPLC
RUO – Research Use Only