N,N-Dimethylacetamide, 99.8+%, for HPLC, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.8+%, for HPLC, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.8+%, for HPLC, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.8+%, for HPLC, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR390670010
Price (MYR)
899.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
899.00
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylacetamide
Melting Point-20.0°C
Absorbance0.046 at 310nm, 0.301 at 285nm, 0.7 (1 cm cell vs HPLC−grade water) at 275nm
CAS127-19-5
ColorColorless
View more
For HPLC
RUO – Research Use Only