tert-Butyl methyl ether, for analysis, Thermo Scientific Chemicals
tert-Butyl methyl ether, for analysis, Thermo Scientific Chemicals
tert-Butyl methyl ether, for analysis, Thermo Scientific Chemicals
tert-Butyl methyl ether, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl methyl ether, for analysis, Thermo Scientific Chemicals
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR390480010
Price (MYR)
508.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
508.00
Add to cart
Chemical Identifiers
CAS708-06-5
IUPAC Name2-hydroxynaphthalene-1-carbaldehyde
Molecular FormulaC11H8O2
InChI KeyNTCCNERMXRIPTR-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Moist powder
Melting point>=77°C (on dry basis)
Appearance (Color)Yellow to brown
HPLC>=93 % (on dry basis)