Iron(III) phosphate hydrate, extra pure
Iron(III) phosphate hydrate, extra pure
Iron(III) phosphate hydrate, extra pure
Iron(III) phosphate hydrate, extra pure
Thermo Scientific Chemicals
Iron(III) phosphate hydrate, extra pure
CAS: 51833-68-2 | FeH2O5P | 168.83 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR389955000500 gGlass bottle
Catalog number ACR389955000
Price (MYR)
939.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
939.00
Add to cart
Chemical Identifiers
CAS35387-93-0
IUPAC Namemethyl 3-iodo-4-methoxybenzoate
Molecular FormulaC9H9IO3
InChI KeyGHNGBFHLUOJHKP-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(OC)C(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Melting point91°C to 95°C