Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
CAS: 124-64-1 | C4H12ClO4P | 190.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR389448000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
800 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS124-64-1, 7732-18-5
Specifications
Chemical Name or MaterialTetrakis(hydroxymethyl)phosphonium chloride
CAS124-64-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Suspected of damaging the unborn child.
Very toxic to aquatic life with lon
CAS7732-18-5
View more
for C-C Bond Formation (Olefination)
RUO – Research Use Only