n-Heptane, 99.5%, for analysis, Thermo Scientific Chemicals
n-Heptane, 99.5%, for analysis, Thermo Scientific Chemicals
n-Heptane, 99.5%, for analysis, Thermo Scientific Chemicals
n-Heptane, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptane, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number ACR389170025
Price (MYR)
1,179.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,179.00
Add to cart
Chemical Identifiers
CAS64584-92-5
IUPAC Namepent-4-en-2-ol
Molecular FormulaC5H10O
InChI KeyZHZCYWWNFQUZOR-UHFFFAOYNA-N
SMILESCC(O)CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
Optical Rotation-4.7 ± 0.1? (neat)
Refractive Index1.4205-1.4265 @ 20?C
For analysis
RUO – Research Use Only