n-Pentane, 99+%, Thermo Scientific Chemicals
n-Pentane, 99+%, Thermo Scientific Chemicals
n-Pentane, 99+%, Thermo Scientific Chemicals
n-Pentane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Pentane, 99+%, Thermo Scientific Chemicals
CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR389070025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1453-82-3
IUPAC Namepyridine-4-carboxamide
Molecular FormulaC6H6N2O
InChI KeyVFQXVTODMYMSMJ-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)153.0-160.0?C
For HPLC
RUO – Research Use Only