Phosphoric acid, pure, Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, pure, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR389021000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6874-60-8
IUPAC Namedecylphosphonic acid
Molecular FormulaC10H23O3P
InChI KeyDZQISOJKASMITI-UHFFFAOYSA-N
SMILESCCCCCCCCCCP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5%
FormCrystals or powder or crystalline powder or waxy solid
Melting Point (clear melt)98.0-104.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White