D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals
CAS: 50-99-7 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number ACR388190010
Price (MYR)
551.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
551.00
Add to cart
Chemical Identifiers
CAS5241-58-7
IUPAC Name(2S)-2-amino-3-phenylpropanamide
Molecular FormulaC9H12N2O
InChI KeyOBSIQMZKFXFYLV-QMMMGPOBSA-N
SMILESN[C@@H](CC1=CC=CC=C1)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5%