Hydrochloric acid, for analysis, 25% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, for analysis, 25% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, for analysis, 25% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, for analysis, 25% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, for analysis, 25% solution in water, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3878000101 LPlastic bottle
Catalog number ACR387800010
Price (MYR)
394.00
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
394.00
Add to cart
Chemical Identifiers
CAS216394-05-7
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Assay (Titration ex Chloride)≥92.0 to ≤108.0%
Appearance (Color)White to cream to yellow
Melting Point65-76°C