Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
CAS: 82113-65-3 | C2HF6NO4S2 | 281.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR387600250
Price (MYR)
3,545.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,545.00
Add to cart
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Coupling reagent
RUO – Research Use Only