Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
CAS: 82113-65-3 | C2HF6NO4S2 | 281.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number ACR387600050
Price (MYR)
1,070.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,070.00
Add to cart
Chemical Identifiers
CAS110-12-3
IUPAC Name5-methylhexan-2-one
Molecular FormulaC7H14O
InChI KeyFFWSICBKRCICMR-UHFFFAOYSA-N
SMILESCC(C)CCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Refractive Index1.4060-1.4090 @ 20°C
FormLiquid
Coupling reagent
RUO – Research Use Only