Pyridine, 99+%, for spectroscopy, Thermo Scientific Chemicals
Pyridine, 99+%, for spectroscopy, Thermo Scientific Chemicals
Pyridine, 99+%, for spectroscopy, Thermo Scientific Chemicals
Pyridine, 99+%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99+%, for spectroscopy, Thermo Scientific Chemicals
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass Bottle
Catalog number ACR386560250
Price (MYR)
485.00
Add to cart
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
485.00
Add to cart
Chemical Identifiers
CAS59843-63-9
IUPAC Name5-(2-methoxyphenyl)-1H-pyrazole
Molecular FormulaC10H10N2O
InChI KeyKLPGJCMICASHKV-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C1=CC=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥96.0%
Appearance (Color)Pale cream
Melting Point (clear melt)80.0-86.0?C
For UV/VIS spectroscopy
RUO – Research Use Only