Boiling stone
Thermo Scientific Chemicals
Boiling stone
For preparative work
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number ACR386212500
Price (MYR)
536.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
536.00
Add to cart
Chemical Identifiers
CAS125-20-2
IUPAC Name3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC28H30O4
InChI KeyLDKDGDIWEUUXSH-UHFFFAOYSA-N
SMILESCC(C)C1=CC(=C(C)C=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Color ChangeColourless @ pH9.3 to blue @ pH10.5
Specific Absorbance (E 1%/1cm)> 750 @ Lambda-max
Formpowder
lambda-max (UV-Vis)590 - 598nm (0.1M NaOH)
Appearance (Color)White