Glass Wool
Thermo Scientific Chemicals
Glass Wool
Filter aid
Have Questions?
Change viewbuttonViewtableView
Quantity:
250g
50g
1kg
Catalog number ACR386062500
Price (MYR)
502.00
Add to cart
Quantity:
250g
Request bulk or custom format
Price (MYR)
502.00
Add to cart
Chemical Identifiers
CAS69676-65-9
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)127.0-136.0?C