Glass Wool
Thermo Scientific Chemicals
Glass Wool
Filter aid
Have Questions?
Change viewbuttonViewtableView
Quantity:
50g
250g
1kg
Catalog number ACR386060500
Price (MYR)
390.00
Add to cart
Quantity:
50g
Request bulk or custom format
Price (MYR)
390.00
Add to cart
Chemical Identifiers
CAS1119-72-8
IUPAC Name(2E,4E)-hexa-2,4-dienedioic acid
Molecular FormulaC6H6O4
InChI KeyTXXHDPDFNKHHGW-ZPUQHVIOSA-N
SMILESOC(=O)C=CC=CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to light beige
Appearance (Form)Crystalline powder
HPLC>=97.5 %