Glass Wool
Thermo Scientific Chemicals
Glass Wool
Filter aid
Have Questions?
Change viewbuttonViewtableView
Quantity:
1kg
50g
250g
Catalog number ACR386060010
Price (MYR)
1,191.00
Add to cart
Quantity:
1kg
Request bulk or custom format
Price (MYR)
1,191.00
Add to cart
Specifications
Quantity1kg
Boiling Point1000°C
Product TypeGlass Wool
For Use With (Application)Filter aid
MaterialGlass