(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
CAS: 18107-18-1 | C4H10N2Si | 114.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
25 mL
Packaging:
Glass bottle
Catalog number ACR385331000
Price (MYR)
4,255.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,255.00
Add to cart
Chemical Identifiers
CAS1303-86-2
IUPAC Nameoxo[(oxoboranyl)oxy]borane
Molecular FormulaB2O3
InChI KeyJKWMSGQKBLHBQQ-UHFFFAOYSA-N
SMILESO=BOB=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Color)White
Total trace metal impurities=<0.2 %
Appearance (Form)Powder
Impurity=<0.01 % (Total halides impurities)
Alkylation reagent
RUO – Research Use Only