(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
CAS: 18107-18-1 | C4H10N2Si | 114.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR385330250
Price (MYR)
1,746.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,746.00
Add to cart
Specifications
Chemical Name or Material(Trimethylsilyl)diazomethane
CAS18107-18-1
CAS Min %32.15
Health Hazard 1GHS Signal Word: Danger
CAS Max %35.54
View more
Alkylation reagent
RUO – Research Use Only