(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes, Thermo Scientific Chemicals
CAS: 18107-18-1 | C4H10N2Si | 114.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR385330050
Price (MYR)
663.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
663.00
Add to cart
Chemical Identifiers
CAS1121-86-4
IUPAC Name1-fluoro-3-iodobenzene
Molecular FormulaC6H4FI
InChI KeyVSKSBSORLCDRHS-UHFFFAOYSA-N
SMILESFC1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
StabilizerCopper powder
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.5%
Refractive Index1.5825-1.5860 @ 20?C
View more
Alkylation reagent
RUO – Research Use Only