Isopropanol, 99.5%, for HPLC gradient grade, Thermo Scientific Chemicals
Isopropanol, 99.5%, for HPLC gradient grade, Thermo Scientific Chemicals
Isopropanol, 99.5%, for HPLC gradient grade, Thermo Scientific Chemicals
Isopropanol, 99.5%, for HPLC gradient grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, 99.5%, for HPLC gradient grade, Thermo Scientific Chemicals
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number ACR383920025
Price (MYR)
1,313.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,313.00
Add to cart
Chemical Identifiers
CAS81-33-4
IUPAC Name7,18-diazaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular FormulaC24H10N2O4
InChI KeyKJOLVZJFMDVPGB-UHFFFAOYSA-N
SMILESO=C1NC(=O)C2=CC=C3C4=CC=C5C(=O)NC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Purple red
Assay from Supplier's CofA≥94%
FormPowder
For HPLC
RUO – Research Use Only