n-Pentane, for analysis, Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR383900025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS481-42-5
IUPAC Name5-hydroxy-2-methyl-1,4-dihydronaphthalene-1,4-dione
Molecular FormulaC11H8O3
InChI KeyVCMMXZQDRFWYSE-UHFFFAOYSA-N
SMILESCC1=CC(=O)C2=C(O)C=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Appearance (Form)Crystalline powder or crystals
Melting point75°C to 78°C
Infrared spectrumConforms
GC>=99.0 %
For analysis
RUO – Research Use Only