n-Pentane, for analysis, Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR383900010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS89-40-7
IUPAC Name5-nitro-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC8H4N2O4
InChI KeyANYWGXDASKQYAD-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2C(=O)NC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point198-204°C
Assay (GC)≥97.5%
For analysis
RUO – Research Use Only