Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals
Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals
Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals
Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
Catalog number ACR383780250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS34725-61-6
IUPAC Namesodium 3,5-dibromo-4-[3-(2,6-dibromo-4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular FormulaC19H9Br4NaO5S
InChI KeyKGANHHDZDYZJEM-UHFFFAOYNA-M
SMILES[Na+].OC1=CC(Br)=C(C(Br)=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=C(Br)C=C([O-])C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
CommentVisual transition interval pH 3.0 (yellow) to pH 4.6 (blue)
CommentClarity of solution P.T.