Chloroform, 99.8%, for analysis, stabilized with amylene, Thermo Scientific Chemicals
Chloroform, 99.8%, for analysis, stabilized with amylene, Thermo Scientific Chemicals
Chloroform, 99.8%, for analysis, stabilized with amylene, Thermo Scientific Chemicals
Chloroform, 99.8%, for analysis, stabilized with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, 99.8%, for analysis, stabilized with amylene, Thermo Scientific Chemicals
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number ACR383770025
Price (MYR)
907.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
907.00
Add to cart
Chemical Identifiers
CAS7417-21-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or pale cream
FormPowder or chunks
Melting Point (clear melt)44.0-50.0?C
Assay (GC)≥97.5%
For analysis
RUO – Research Use Only