Sodium hydroxide, for analysis, micropearls, Thermo Scientific Chemicals
Sodium hydroxide, for analysis, micropearls, Thermo Scientific Chemicals
Sodium hydroxide, for analysis, micropearls, Thermo Scientific Chemicals
Sodium hydroxide, for analysis, micropearls, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, for analysis, micropearls, Thermo Scientific Chemicals
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number ACR383040010
Price (MYR)
620.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
620.00
Add to cart
Chemical Identifiers
CAS123-51-3
IUPAC Name3-methylbutan-1-ol
Molecular FormulaC5H12O
InChI KeyPHTQWCKDNZKARW-UHFFFAOYSA-N
SMILESCC(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Color scale=<20 APHA
Refractive index1.4050 to 1.4070 (20°C, 589 nm)
GC>=98.5 %