4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
CAS: 3731-53-1 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ACR382081000
Price (MYR)
1,104.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,104.00
Add to cart
Chemical Identifiers
CAS33626-98-1
IUPAC Namemethyl 6-bromonaphthalene-2-carboxylate
Molecular FormulaC12H9BrO2
InChI KeyJEUBRLPXJZOGPX-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C2C=C(Br)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to cream to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Melting point118°C to 126°C