4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%, Thermo Scientific Chemicals
CAS: 3731-53-1 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ACR382080250
Price (MYR)
496.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
496.00
Add to cart
Chemical Identifiers
CAS14107-97-2
IUPAC Name1,3,5-trimethoxy-2-methylbenzene
Molecular FormulaC10H14O3
InChI KeyTZPKFPYZCMHDHL-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=C(C)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to yellow
Refractive Index1.5285-1.5335 @ 20?C (as melt)
FormLow melting crystals or powder or crystalline powder or clear liquid
Assay (GC)≥96.0%