Methyl 3-nitrosalicylate, 98%, Thermo Scientific Chemicals
Methyl 3-nitrosalicylate, 98%, Thermo Scientific Chemicals
Methyl 3-nitrosalicylate, 98%, Thermo Scientific Chemicals
Methyl 3-nitrosalicylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl 3-nitrosalicylate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR381920050
Price (MYR)
1,515.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,515.00
Add to cart
Chemical Identifiers
CAS6389-79-3
IUPAC Namemethyl methyl(phenyl)phosphinate
Molecular FormulaC8H11O2P
InChI KeyRTAAKNLNWIHGGX-UHFFFAOYNA-N
SMILESCOP(C)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5260-1.5300 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%