Piperine, 98%, Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
CAS: 94-62-2 | C17H19NO3 | 285.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR381450050
Price (MYR)
549.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
549.00
Add to cart
Specifications
Chemical Name or MaterialPiperine
CAS94-62-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.