Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
CAS: 96-10-6 | C4H10AlCl | 120.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR381151000
Price (MYR)
743.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
743.00
Add to cart
Chemical Identifiers
CAS13329-40-3
IUPAC Name1-(4-iodophenyl)ethan-1-one
Molecular FormulaC8H7IO
InChI KeyJZJWCDQGIPQBAO-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Crystalline powder
GC>=97.5 %
Melting point82°C to 89°C
Infrared spectrumConforms
Lewis acid
RUO – Research Use Only