Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
CAS: 90076-65-6 | C2F6LiNO4S2 | 287.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number ACR381030500
Price (MYR)
2,728.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,728.00
Add to cart
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.95% (metals basis)
FormSlug, 6.35mm (0.25in) diameter.