Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%, Thermo Scientific Chemicals
CAS: 90076-65-6 | C2F6LiNO4S2 | 287.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number ACR381030100
Price (MYR)
966.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
966.00
Add to cart
Chemical Identifiers
CAS1302-42-7
IUPAC Namesodium dioxoalumanuide
Molecular FormulaAlNaO2
InChI KeyIYJYQHRNMMNLRH-UHFFFAOYSA-N
SMILES[Na+].O=[Al-]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble material1% max