4-Ethynylanisole, 97%, Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
CAS: 768-60-5 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR379920050
Price (MYR)
1,356.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,356.00
Add to cart
Chemical Identifiers
CAS2373-79-7
IUPAC Name(2E)-3-(4-ethoxyphenyl)prop-2-enoate
Molecular FormulaC11H11O3
InChI KeyDZLOUWYGNATKKZ-VMPITWQZSA-M
SMILESCCOC1=CC=C(C=CC([O-])=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Melting Point (clear melt)194.0-200.0?C
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.0%