4-Ethynylanisole, 97%, Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Ethynylanisole, 97%, Thermo Scientific Chemicals
CAS: 768-60-5 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR379920010
Price (MYR)
574.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
574.00
Add to cart
Chemical Identifiers
CAS61970-08-9
IUPAC Name4-{6-[(1-hydroxy-3-methylbutan-2-yl)amino]imidazo[1,2-b]pyridazin-3-yl}benzonitrile
Molecular FormulaC18H19N5O
InChI KeyCQZVJEKKNHOFNB-UHFFFAOYNA-N
SMILESCC(C)C(CO)NC1=NN2C(C=C1)=NC=C2C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormSpherical microparticle beads
CommentSuspension in 20% ethanol as antimicrobial agent
Particle SizeVolume share for range 45-165µm: ≥95%
Appearance (Color)White