Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals
Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals
Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals
Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals
CAS: 2567-83-1 | C8H20ClNO4 | 229.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Plastic bottle
Catalog number ACR379801000
Price (MYR)
403.00
Add to cart
Quantity:
100 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
403.00
Add to cart
Chemical Identifiers
CAS2567-83-1, 7732-18-5
Specifications
Chemical Name or MaterialTetraethylammonium perchlorate
CAS2567-83-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May intensify fire; oxidizer.
Risk of explosion if heated under confinement.
CAS7732-18-5
View more
Oxidation reagent, Phase transfer catalyst
RUO – Research Use Only