tert-Butyl methyl ether, 99.9%, extra pure, Thermo Scientific Chemicals
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change view
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR378720010
Price (MYR)
504.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
504.00
Add to cart
Specifications
Chemical Name or Materialtert-Butyl methyl ether
Melting Point-110.0°C
CAS1634-04-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS P Statement Causes skin irritation. Highly flammable liquid and vapor.