N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR378710025
Price (MYR)
955.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
955.00
Add to cart
Chemical Identifiers
CAS429-42-5
IUPAC Nametetrabutylazanium; tetrafluoroboranuide
Molecular FormulaC16H36BF4N
InChI KeyNNZZSJSQYOFZAM-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=97.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder