N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR378710010
Price (MYR)
573.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
573.00
Add to cart
Chemical Identifiers
CAS1188-14-3
IUPAC Nametriethylgermane
Molecular FormulaC6H16Ge
InChI KeyLTLGQZPDSQYFSB-UHFFFAOYSA-N
SMILESCC[GeH](CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥96.0%