Calcium phosphate, monobasic, 90%, pure
Calcium phosphate, monobasic, 90%, pure
Calcium phosphate, monobasic, 90%, pure
Calcium phosphate, monobasic, 90%, pure
Thermo Scientific Chemicals
Calcium phosphate, monobasic, 90%, pure
CAS: 7758-23-8 | CaH4O8P2 | 234.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3786100101 kgPlastic bottle
Catalog number ACR378610010
Price (MYR)
751.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
751.00
Add to cart
Chemical Identifiers
CAS123-35-3
IUPAC Name7-methyl-3-methylideneocta-1,6-diene
Molecular FormulaC10H16
InChI KeyUAHWPYUMFXYFJY-UHFFFAOYSA-N
SMILESCC(C)=CCCC(=C)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4660 to 1.4720 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=88 %