Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number ACR378418000
Price (MYR)
3,314.00
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
3,314.00
Add to cart
Chemical Identifiers
CAS74-88-4, 1634-04-4
Specifications
Chemical Name or MaterialIodomethane
Name Note2M Solution in tert-Butyl Ether
CAS74-88-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Highly flammable liquid and vapor.
View more
Alkylation reagent
RUO – Research Use Only