Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodomethane, 2M solution in tert-Butyl methyl ether, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number ACR378411000
Price (MYR)
710.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
710.00
Add to cart
Chemical Identifiers
CAS563-46-2
IUPAC Name2-methylbut-1-ene
Molecular FormulaC5H10
InChI KeyMHNNAWXXUZQSNM-UHFFFAOYSA-N
SMILESCCC(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
CommentPurchased in U.K. and U.S.
Assay (GC)≥97.5% (U.K. specification)
Appearance (Color)Clear colorless to pale yellow
Assay from Suppliers CofA≥97.5% (U.S. specification)
Refractive Index1.3735-1.3785 @ 20°C (non-U.S. specification)
View more
Alkylation reagent
RUO – Research Use Only