Potassium thiocyanate, 98%, pure, Thermo Scientific Chemicals
Potassium thiocyanate, 98%, pure, Thermo Scientific Chemicals
Potassium thiocyanate, 98%, pure, Thermo Scientific Chemicals
Potassium thiocyanate, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium thiocyanate, 98%, pure, Thermo Scientific Chemicals
CAS: 333-20-0 | CKNS | 97.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR377435000
Price (MYR)
695.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
695.00
Add to cart
Chemical Identifiers
CAS14548-48-2
IUPAC Name4-(4-chlorobenzoyl)pyridine
Molecular FormulaC12H8ClNO
InChI KeyYMTFKKLFLLAFNI-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)C(=O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥99.0%
Melting Point (clear melt)104.0-108.0?C
Appearance (Color)Cream
FormPowder
Indicator
RUO – Research Use Only