Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals
Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals
Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals
Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals
CAS: 128446-36-6 | C54H94O35 | 1303.311 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR377110250
Price (MYR)
986.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
986.00
Add to cart
Chemical Identifiers
CAS80-08-0
IUPAC Name4-(4-aminobenzenesulfonyl)aniline
Molecular FormulaC12H12N2O2S
InChI KeyMQJKPEGWNLWLTK-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms (non-U.S. specification)
Melting Point (clear melt)175.0-181.0°C (non-U.S. specification)
Chelation/Complexation reagent
RUO – Research Use Only